Synthware Stopcock, 1:10 Glass Plug, Straight Bore, Interchangeable, Kemtech America
Supplier: Kemtech America
S321230 S321740 S322044
60005-156EA 92.19 CAD
60005-156 60005-158 60005-160
Synthware Stopcock, 1:10 Glass Plug, Straight Bore, Interchangeable, Kemtech America
Stopcocks
Stopcock with 1:10 taper straight bore glass plug
  • Heavy wall borosilicate construction
  • Hose connection on both sides
Plugs and barrels are ground to be interchangeable.