SPILFYTER® Hazmat Pad, NPS
Supplier: NPS
S2-75 S2-70 S2-71
15551-702CS 597.94 CAD
15551-702 56609-392 56609-396
SPILFYTER® Hazmat Pad, NPS
Spill Control Materials
Single-weight perforated pad absorbs up to 16 times its own weight in virtually all liquids.
  • Absorbs up to 16X its own weight in virtually all liquids, aggressive or nonaggressive
  • Perforated, allows quick response when nature of spill is unknown
  • Pad is constructed of smaller diameter fibers providing greater liquid retention and strength
  • Yellow color alerts others of potential danger
Single-weight perforated pad absorbs up to 16 times its own weight in virtually all liquids, either aggressive or nonaggressive, and allows a quick response when the nature of the spill is unknown. Pad is constructed of smaller diameter fibers that provide greater liquid retention and strength. Yellow color alerts others of potential danger.