Synthware Sublimators, Kemtech America
Supplier: Kemtech America
S732510 S734040 S735060
60004-068EA 552.57 CAD
60004-068 60004-070 60004-072
Synthware Sublimators, Kemtech America
Condensers
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and a lower vessel to hold the material to be sublimed.
  • Heavy wall Borosilicate construction
  • Horseshoe clamp included
  • Grease free
  • Hi-Vac Valve included
The sublimate collects on the lower end of the condenser, which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. The condenser and the lower vessel are connected by an O-ring joint, which enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring, and stainless steel pinch clamp.