SPILFYTER® Very Low Lint Sorbents, NPS
Supplier: NPS
SPILFYTER®
225121 225122 225112
56609-194CS 997.03 CAD
56609-194 56616-356 56616-350
SPILFYTER® Very Low Lint Sorbents, NPS
Spill Control Materials
Made from virtually lint-free and nonbiodegradable universal polypropylene material
Material is soft, flexible, and tough. Wringing removes chemicals collected (chemical-resistant gloves must be worn). Sheets and rolls are constructed of one-ply, heat-pressed, treated polypropylene. Pads are made of several layers of polypropylene sheets that are heat-pressed in one-ply heat-sealed polypropylene pouches. Sorbents are polybagged and heat-sealed to prevent contamination.
Sorbent sheets, rolls, and pads soak up eight to 10 times their weight in chemicals, oils, solvents, and emulsions without significant particulate loss.