SPILFYTER® Sorbents for Hydrocarbons, NPS
Supplier: NPS
M-75 M-37 M-55
15551-692CS 365.14 CAD
15551-692 15551-746 15551-880
SPILFYTER® Sorbents for Hydrocarbons, NPS
Spill Control Materials
For quick absorption of hydrocarbon spills
Sock is designed for quick, easy oil pick-up. The superior absorbency and wicking action make this sock especially suitable for recovering oil spills on land.
Melt-blown pad is ideal for collection of lightweight hydrocarbons such as J P-4 fuel and kerosene.
Boom is designed to handle easily on water. Absorbs oils quickly. The sealed polypropylene case makes the boom environmentally safe.
SPILFYTER® Sorbents are made of 100% polypropylene fabric that repels water while attracting and absorbing oils and hydrocarbon-based fuels.
Order Now
SPECIFICATIONS
Environmentally Preferable