Synthware Sublimation Apparatuses with High Vacuum Valve, Kemtech America
Supplier: Kemtech America
S740100 S740150
60004-086EA 945.4 CAD
60004-086 60004-088
Synthware Sublimation Apparatuses with High Vacuum Valve, Kemtech America
Condensers
A grease-free sublimer with a flat bottom to enable heating on a hot plate stirrer.
  • Heavy wall Borosilicate construction
  • Horseshoe clamp included
  • Grease free
  • Hi-Vac Valve included
Condenser has a large top opening for use with dry ice, acetone, or liquid nitrogen. The flask and condenser are connected by a flat O-ring joint, permitting parts to be gently separated for maximum product recovery. Vacuum control is provided by a 0–8 mm high vacuum valve. Complete apparatus consists of the cold finger condenser, flask, Viton® O-ring, and horseshoe clamp.More Product Information