Coliwasa Samplers, PVC, Reusable, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
885250 885252 885253
66174-250EA 1011.54 CAD
66174-250 66174-252 66174-253
Coliwasa Samplers, PVC, Reusable, Wheaton®, DWK Life Sciences
Samplers Fluid Samplers
For safe and simple sampling of drums and tanks.
Uniquely designed top spin bar and bottom valve plate mechanism produce a positive seal for sample retention. Provides excellent control for releasing the sample into containers. Modular design adds flexibility by allowing the connection of several samplers to form a sampling unit up to 20 feet in length via connectors (sold separately). Samplers are chemically-resistant, gray colored PVC.
Ordering information: Samplers and connectors (sold separately) available in 121.9 cm (48") and 182.9 cm (72") lengths. Samplers and connectors each supplied with a 121.9 cm (48") long cleaning brush.