Saline Solutions, NERL Diagnostics
Supplier: Thermo Fisher Scientific
850510L 8504
CA48212-271LEA 562.4 CAD
CA48212-271L 72060-035
Saline Solutions, NERL Diagnostics
Buffers Cell Culture Buffers
Blood bank saline pH 7.0–7.2 is a phosphate buffered saline solution manufactured to match the pH of human blood
Manufactured to meet NCCLS standard ASI/BB-1. Certificates of analysis are available.
Note: Not for infusion, irrigation, or injection.
This preservative-free sodium chloride solution is for use in serological (blood bank) testing or for general laboratory use.