Tissue Powders for Adsorption, Rockland Immunochemicals
Supplier: Rockland Immunochemical
C101 C102 C103 C113 C115 C116 C117 C119 C120 C121 C122 C123 C124
CAC100EA 747.43 CAD
CAC100 CAC101 CAC102 CAC103 CAC104 CAC105 CAC106 CAC107 CAC108 CAC109 CAC110 CAC111 CAC112 CAC113 CAC114 CAC115 CAC116 CAC117 CAC118 CAC119 CAC120 CAC121 CAC122 CAC123 CAC124
Tissue Powders for Adsorption, Rockland Immunochemicals
Media for Microbiology Dehydrated Media for Microbiology
Use for protocols that require a high nuclear protein content
Powders are produced by washing the various organs repeatedly in saline to remove blood, precipitated in acetone, and micronized to a fine powder. Keep container sealed and store at 2–8°C (36–46°F). All vials contain 1g of powder.
Order Now
SPECIFICATIONS
Environmentally Preferable