Exit Signs, Brady®
Supplier: Brady Worldwide
37851 38097 38098 90839 90840 90841 90842 90885 90887
CA89007-140EA 343.61 CAD
CA89007-140 CA89007-142 CA89007-144 CA89007-874 CA89007-876 CA89007-878 CA89007-880 CA89007-898 CA89007-902
Exit Signs, Brady®
Signs
Photoluminescent exit signs are steel-encased glow-in-the-dark plastic panels
Three types of framed exit signs are available: single-face wall-mounted, single-face with mounting bracket, or double-face with mounting bracket. Signs come in three colors: black, red, and green. Extra brackets are also available.
These signs provide consistent and highly visible illumination immediately after the lights go out. Signs absorb and store energy from ambient light. They use zero electricity and do not require special wiring, maintenance, a backup power source, or professional installation. Punch-out or field-applied chevrons provide direction. Signs are easily installed and are perfect for high and low level applications. Intended for indoor use only. Compliant with all current building codes.