Samco™ UrineGUARD™ Urine Collection Containers, Thermo Scientific
Supplier: Thermo Scientific
UrineGUARD®
30 2500
14005-980CS 864.54 CAD
14005-980
Samco™ UrineGUARD™ Urine Collection Containers, Thermo Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Alleviate mess and exposure when handling urine collection and sampling with the Thermo Scientific Samco 24-hour Urine Collection product line
These HDPE containers are amber tinted to protect from light exposure and are inert to normal preservatives.