Arachidonic acid (from porcine liver), Calbiochem®
Supplier: MilliporeSigma
181198-100MG
181198-1GM
CA80050-322EA
404.71
CAD
CA80050-322
CA80050-324
Arachidonic acid (from porcine liver), Calbiochem®
Arachidonic acid
Precursor for prostaglandins, prostacyclin and thromboxane
Binds to G-protein [alpha]-subunits in a covalent, post-translational manner. Inhibits Ras-GAP. Stimulates the synthesis of nitric oxide by platelets. Modulates nitric oxide production by prostaglandin synthesis via the cyclooxygenase pathway.
Formula:
C₂₀H₃₂O₂ |
MDL Number:
MFCD00004417 CAS Number: 506-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...