PIG® Mat Tablet Pack, New Pig
Supplier: New Pig
MAT127 MAT124
56620-554PK 299.49 CAD
56620-554 56620-576
PIG® Mat Tablet Pack, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated
Absorbs and retains oils, coolants, solvents and water. Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. Pop-up tabs and cardboard back let you hang pack anywhere in your plant - on a pegboard by your workbench, on your tool cart, even on your forklift. Fixed number of pads makes it easy to monitor inventory and usage. Perforated at the bound edge - one quick tear is all it takes and you're ready to work.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. Light-weight construction is ideal for small tasks and cleanups.
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind