Collector Vials, Starplex®
Supplier: Starplex
V8443 V302 V302-F V302-SH V9581 B300-NLS V302-PIN V9487 V302-NL
CA68810-122CS 382.56 CAD
CA68810-122 CA73240-135 CA73240-137 CA73240-145 CA68810-124 CA73240-140 CA11008-312 CA73240-139L CA10161-220 CA10510-908
Collector Vials, Starplex®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
These 30mL vials are manufactured using break-resistant, clarified polypropylene, to ensure the consistency and reliability of the product
The universal collector is sterile and can be used for multiple purposes including fecal, urine, and sputum collection. The vial has a label and easy-to-read graduations, which assist in the collection of the proper sample size.
The pinworm collector features a clear polystyrene paddle insert cap, lightly adhesive on one side allowing easy collection of ova. The vial has a label and easy-to-read graduations.
The fecal collector features aMore Product Information