Shipping Labels, DOT, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
DL53AL DL25AL DL26AL DL43AL
56606-660PK 216.87 CAD
56606-660 56606-662 56606-663 56606-669
Shipping Labels, DOT, National Marker
Labels
Pressure-sensitive paper labels comply with DOT HM-181 specifications when shipping hazardous materials (DOT 49 CFR 172.407).
Ordering information: Also available as 0.004 mm pressure-sensitive vinyl labels; contact your Avantor representative for details.