PIG® Original Absorbent Sock, New Pig
Supplier: New Pig
404 PIG218 204 PIG209
46000-520PK 224.85 CAD
46000-520 46000-522 56620-586 56620-692
PIG® Original Absorbent Sock, New Pig
Spill Control Materials
Formable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and help keep workers safe.
  • Absorbs and retains oils, coolants, solvents and water
  • Corncob filler is light-weight and economical to handle general, everyday tasks
  • Can be incinerated after use to reduce waste or for fuels blending
Contained corncob filler pulls liquids from the floor for maximum absorbency while making cleanup quick and easy. Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated.