1-Octanesulfonic acid sodium salt monohydrate ≥99% for HPLC, for ion pair chromatography, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sodium 1-octansulphonate monohydrate, Sodium 1-octansulfonate monohydrate, Sodium 1-octanesulphonate monohydrate

OX0101-3 OX0101-4 OX0101-2
CAOX0101-3EA 1273 CAD
CAOX0101-3 CAOX0101-4 CAOX0101-2
1-Octanesulfonic acid sodium salt monohydrate ≥99% for HPLC, for ion pair chromatography, Sigma-Aldrich®
1-Octanesulphonic acid sodium salt monohydrate

Used to separate pharmaceutical compounds from their metabolites


Compounds can be separated on a reversed phase column with the addition of an ion pairing reagent to the mobile phase. The type of ion pairing reagent and its concentration give different selectivities.

Formula: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 207596-29-0

Order Now

Specification Test Results

Color White
Form Solid
Purity 99% min
Solubility in water (5% w/v) To pass test
UV Abs. at 210 nm 1.00AU max
UV Abs. at 220 nm 0.10AU max
UV Abs. at 254 nm 0.06AU max
UV Abs. at 280 nm 0.06AU max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR