Sani-S'wipe Sterile Sampling Kits
Supplier: Labplas
KSS-68230 KSS-68230-BPW-100 KSS-68230-NB-100
77531-608CS 331.31 CAD
77531-608 77531-610 77531-612
Sani-S'wipe Sterile Sampling Kits
Samplers Disposable Samplers
The Sani-S'wipe is a kit comprising a blue cloth and a sterile bag.
  • ISO 18593 compliant
  • Time-saving solution
  • Ideal for various surfaces
  • Certified sterile
  • Integrated closure system
  • Biocide-free cloth for sampling
  • Thermoplastic laminate material
  • Transparent
The Sani-S'wipe is a kit comprising a blue cloth and a sterile bag, designed for the collection of samples used to detect the presence of microbiological contaminants and other foodborne pathogens on various surfaces. It is easy to use and is ideal for large areas. It guarantees an accurateMore Product Information