Crews® Protective Goggles, MCR Safety
Supplier: MCR Safety
2230R 2220R 2225R 2235R 2237R
11000-398CS 2782.21 CAD
11000-398 80081-776 80081-778 80081-782 80081-784
Crews® Protective Goggles, MCR Safety
Goggles
Goggles provide a wide, unobstructed field of vision and are available in a variety of styles to protect against impact and splash.
  • Wide, unobstructed field of vision
  • Can be worn with most half-mask respirators and will fit comfortably over most prescription eyewear
  • Transparent PVC frame and clear 0.06mm thick polycarbonate lens with scratch-resistant Duramass® coating
  • Latex-free rubber straps provide secure fit on all styles
  • Lenses provide 99.9% UV protection
  • Goggles meet or exceed ANSI Z87.1+ safety standards
All goggles feature a transparent PVC frame and clear 0.06mm thick polycarbonate lens with scratch-resistant Duramass® coating. Latex-free rubber straps provide a secure fit on all styles. Lenses provide 99.9% UV protection.
They can be worn with most half-mask respirators and will fit comfortably over most prescription eyewear.
Certifications: Goggles meet or exceed ANSI Z87.1+ safety standards.