Calbiochem® Proteasome inhibitor II
Supplier: MilliporeSigma
539162-1MG 539162-5MG
CA80053-878EA 369.29 CAD
CA80053-878 CA80053-880
Calbiochem® Proteasome inhibitor II
Proteins and Peptides
A potent and cell-permeable proteasome inhibitor
Inhibits the chymotrypsin-like activity of the pituitary multicatalytic proteinase complex (MPC; Ki=460nM). Does not inhibit the peptidylglutamyl-peptide hydrolyzing activity of MPC even at concentrations of 200µM. Also blocks the decay of l[kappa]B-[alpha] and l[kappa]B-[beta] proteins in exponentially growing WEHI 231 cells.