Calbiochem® MG-132
Supplier: MilliporeSigma
474790-1MG 474790-5MG
CA80053-194EA 168.33 CAD
CA80053-194 CA80053-196
Calbiochem® MG-132
Proteins and Peptides
A potent, reversible, and cell-permeable proteasome inhibitor (Ki=4nM)
Reduces the degradation of ubiquitin-conjugated proteins in mammalian cells and permeable strains of yeast by the 26S complex without affecting its ATPase or isopeptidase activities. Activates c-Jun N-terminal kinase (JNK1), which initiates apoptosis. Inhibits NF-[kappa]B activation (IC50=3µM).