(R)-Roscovitine, Calbiochem®

Supplier: MilliporeSigma

557360-1MG 557360-5MG
CA80054-314EA 202.45 CAD
CA80054-314 CA80054-316
(R)-Roscovitine, Calbiochem®
(R)-Roscovitine

A potent and selective inhibitor of cyclin-dependent kinases (cdks) that exhibits about ten-fold greater efficacy towards p34cdc2 and p33cdk2, and twenty-fold greater efficacy towards p33cdk5 relative to olomoucine (80053-494, -496)


Inhibits p34cdc2/cyclin B (IC50=650nM), p33cdk2/cyclin A (IC50=700nM), p33cdk2/cyclin E (IC50=200nM), and p33cdk5/p35 (IC50=200nM) by competing for the ATP binding domain of the kinases. Exhibits reduced sensitivity towards related kinases, including ERK1 and ERK2 (IC50=34µM and 14µM respectively). Roscovitine does not significantly affect the activity of other protein kinases even at 100µM, including p34cdk4/cyclin D1 and p40cdk6/cyclin D2. Compared to olomoucine, roscovitine displays increased anti-mitotic activity at the G1/S and G2/M phases of the cell cycle.

Formula: C₁₉H₂₆N₆O
MDL Number: MFCD02266401
CAS Number: 186692-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR