Thermal Transfer Printer Labels, Brady
Supplier: Brady Worldwide
66267 66269
CA89009-232EA 516.5 CAD
CA89009-232 CA89009-234
Thermal Transfer Printer Labels, Brady
Labels
These printable labels allow for application to frozen surfaces such as glass and polypropylene stored in liquid nitrogen.
  • Polyester construction
  • Permanent acrylic adhesive
  • White with matte finish
Designed for general laboratory identification, labels offer excellent print smudge resistance, solvent resistance, and low temperature performance. Labels also perform well in autoclave applications.