Nitro blue tetrazolium chloride (NBT), Calbiochem®

Supplier: MilliporeSigma
Warning

484235-1GM 484235-250MG
CA80057-128EA 546.57 CAD
CA80057-128 CA80057-130
Nitro blue tetrazolium chloride (NBT), Calbiochem®
Nitro blue tetrazolium chloride (NBT)

NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50=3–4µM). A well-known scavenger of superoxide anions. Useful as a substrate for alkaline phosphatase, often in conjunction with BCIP.

Formula: C₄₀H₃₀Cl₂N₁₀O₆
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR