Toluene-α-sulfonyl fluoride ≥98%, Calbiochem®

Supplier: MilliporeSigma
Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulfonyl fluoride, α-Toluenesulphonyl fluoride, Phenylmethanesulfonyl fluoride, Phenylmethylsulfonyl fluoride

52332-1GM 52332-25GM 52332-5GM
CA80055-380EA 173.45 CAD
CA80055-380 CA80055-382 CA80055-384
Toluene-α-sulfonyl fluoride ≥98%, Calbiochem®
Toluene-α-sulphonyl fluoride

Irreversible inhibitor of serine proteases. Its mechanism of action is analogous to that of diisopropylfluorophosphate. PMSF causes sulfonylation of the active-site serine residues. Also reported to inhibit internucleosomal DNA fragmentation in immature thymocytes. Effective concentration: 50µM.

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Boiling Pt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR