Calbiochem® Leupeptin hemisulfate
Supplier: MilliporeSigma
108975-100MG 108975-10MG 108975-25MG 108975-50MG 108975-5MG
CA80050-014EA 2128.91 CAD
CA80050-014 CA80050-016 CA80050-018 CA80050-020 CA80050-022
Calbiochem® Leupeptin hemisulfate
Proteins and Peptides
A reversible inhibitor of trypsin-like proteases and cysteine proteases
Also known to inhibit activation-induced programmed cell death and to restore defective immune responses of HIV+ donors. Effective concentration: 10–100µM.