VWR® Slide Labels
Supplier: VWR International
89097-832 89097-842 89097-844 89097-854
89097-832CS 31.34 CAD
89097-832 89097-842 89097-844 89097-854
VWR® Slide Labels
Labels
Designed to help prevent curling or detaching, slide labels are printed on smudgeproof litho sheets and feature a permanent adhesive.
  • Labels have a thickness 0.012 cm (0.0078")
  • Color: white