Caspase-3 Inhibitor I, Calbiochem®
Supplier: MilliporeSigma
Synonyms: N-Acetyl-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Glu-Val-Asp-al, Ac-A-A-V-A-L-L-P-A-V-L-L-A-L-L-A-P-D-E-V-D-CHO, DEVD-CHO, CPP32/Apopain Inhibitor
235420-1MG 235420-5MG
CA80051-228EA 322.76 CAD
CA80051-228 CA80051-230
Caspase-3 Inhibitor I, Calbiochem®
Caspase-3 Inhibitor I
A very potent, specific, and reversible inhibitor of caspase-3 (IC50=200 pM), caspase-6, caspase-7, caspase-8, and caspase-10.
Sold on the basis of peptide content.