Caspase-1 Inhibitor VI, Calbiochem®

Supplier: MilliporeSigma

218746-1MG 218746-250UG
CA80050-802EA 1385.42 CAD
CA80050-802 CA80050-804
Caspase-1 Inhibitor VI, Calbiochem®
Caspase-1 Inhibitor VI

A potent, cell-permeable, and irreversible inhibitor of caspase-1 (ICE) and caspase-4.

Formula: C₃₁H₃₉FN₄O₉
MDL Number: MFCD06798973
CAS Number: 210344-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR