Calbiochem® Caspase Inhibitor III
Supplier: MilliporeSigma
218745-1MG 218745-250UG
CA80050-798EA 750.98 CAD
CA80050-798 CA80050-800
Calbiochem® Caspase Inhibitor III
Proteins and Peptides
A cell-permeable, irreversible, broad-spectrum caspase inhibitor.