VWR® Write-On/Wipe-Off Magnetic Shelf Label Rolls
Supplier: VWR International
MX-100 MX-200 MX-300
80094-278EA 297.92 CAD
80094-278 80094-280 80094-282
VWR® Write-On/Wipe-Off Magnetic Shelf Label Rolls
Labels
Magnetic label material has a white vinyl surface that may be written on with wet-erase marker and then wiped off.
  • May be cut down to any size
Provided in a rol. Thickness: 0.8 mm (¹/₃₂"). Roll length: 15.2 m (50').