BAPTA-AM, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester

196419-100MG 196419-25MG
CA80050-392EA 1353.48 CAD
CA80050-392 CA80050-394
BAPTA-AM, Calbiochem®
BAPTA-AM

Membrane-permeable form of BAPTA


Can be loaded into a variety of cells, where it is hydrolyzed by cytosolic esterases and is trapped intracellularly as the active chelator BAPTA. Prevents cocaine-induced ventricular fibrillations. Abolishes vitamin D3-induced increase in intracellular Ca2+. Induces activation of protein kinase C. Also inhibits thapsigargin-induced apoptosis in rat thymocytes.

Formula: C₃₄H₄₀N₂O₁₈
MW: 764.69 g/mol
MDL Number: MFCD00036696
CAS Number: 126150-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR