Microliter™ 7000 Series Syringes, Hamilton
Supplier: Hamilton
86211 88411 88511 88011 80100 88400 88500 80135 86259 86250 86257 86200 88000
60374-006EA 291.62 CAD
60374-006 60374-008 60374-010 60374-012 60375-034 60375-067 60375-078 60375-158 60374-004 60375-012 60375-023 60375-045 60375-089
Microliter™ 7000 Series Syringes, Hamilton
Syringes Syringes for Chromatography
7000 Series syringes feature a unique modified Microliter™ design based on the dead volume-free principle of positive displacement
Syringe Termination: A knurled hub is used exclusively on 7000 Series syringes. The hub handles up to 6000psig maximum injection pressure. The needle is removable, but with a limited number of gauges available because the plunger is fitted inside the needle. Autoclavable when disassembled. Gas sterilizable. Repeated autoclaving will shorten syringe life.
Needle Point Styles:
Point style 2 is a sharp, beveled, curved, non-coring needle point recommendedMore Product Information