Calbiochem® ALLN
Supplier: MilliporeSigma
Inhibitor of calpain I (Ki=190nM), calpain II (Ki=220nM), cathepsin B (Ki=150nM) and cathepsin L (Ki=0.5nM). Inhibits neutral cysteine proteases and the proteasome (Ki=6µM). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Also inhibits retrovirus-induced apoptosis in L929 cells. Inhibits the proteolysis of I[kappa]B-[alpha] and I[kappa]B-[beta] by the ubiquitin-proteasome complex. Inhibits cell cycle progression at G1/S and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene.