Mounting Medium, Epredia™
Supplier: RICHARD-ALLAN SCIENTIFIC CA
4111 4112
CA41810-034CS 520.82 CAD
CA41810-034 CA84000-112
Mounting Medium, Epredia™
Stains Microscopy and staining reagents
Store slides for prolonged periods of time without fading of stains with Epredia™ mounting medium.
  • Rapid drying
  • Toluene-based
  • Optically clear
This toluene-based rapid drying mounting medium is miscible with all clearing agents including xylene substitutes. It is optically clear and will not cause fading of stains, even after prolonged storage. This mounting medium is miscible with all clearing agents, including xylene substitutes.
Ordering information: The 118.3 ml (4 oz.) bottle (84000-112) is a squeeze bottle with a twist-open/close cap.
Order Now
SPECIFICATIONS
Environmentally Preferable