Satin Cloth Labels, Shamrock
Supplier: Shamrock Scientific
SC-1 SC-2 SC-3
36436-507PK 408.13 CAD
36436-507 36436-530 36436-562
Satin Cloth Labels, Shamrock
Labels
Ideal for cryogenic procedures that use dry ice, liquid nitrogen, or deep freezing.
Labels withstand extremely cold temperatures. Rayon cloth resists acids, alkalies, oils, grease, soap, and fresh or salt water. Textured surface is easily marked with pen, pencil, or felt-tip marker. Pressure sensitive backing adheres to almost any surface.
Order Now
SPECIFICATIONS
Environmentally Preferable