VWR® SPE Glass Column Processors
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR7208-00 BAKR6998-00
CAJT7208-0EA 6327.05 CAD
CAJT7208-0 CAJT6998-0
VWR® SPE Glass Column Processors
Vacuum Traps
These glass column processors are specially designed glass vacuum chambers that allow multiple samples to be run simultaneously. The glass chamber allows unobstructed viewing of samples at all times.
  • Run multiple samples simultaneously
  • Unobstructed viewing of samples
  • Specially-designed, glass vacuum chambers
The SPE-12G processor allows up to 12 samples to be run simultaneously and includes 12 Luer-type fittings, 12 individual flow control stopcocks, 12 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable