Human Myeloperoxidase (from Polymorphonuclear Leukocytes)
Supplier: MilliporeSigma
475911-100UG 475911-100UG
CA80053-242EA 483.62 CAD
80053-242 CA80053-242
Human Myeloperoxidase (from Polymorphonuclear Leukocytes)
Enzymes
100mM NaCl, 50mM sodium acetate, pH 6.0
A lysosomal heme protein that catalyzes oxidations by hydrogen peroxide, including MPO-chloride-mediated killing of microbes and tumor cells, inactivation of chemotactic factors and cross-linking of proteins. Specific activity: 150–200units/mg protein. Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.