QuEChERS SPE Tubes, Restek
Supplier: Restek
26124 26125 26128 26129 26123 26126 26127
RK26124PK 525.87 CAD
RK26124 RK26125 RK26128 RK26129 RK26123 RK26126 RK26127
QuEChERS SPE Tubes, Restek
Chromatography Columns
These tubes comply with the USDA's QuEChERS method, used in the extraction and clean-up of pesticide residue samples.
  • Contained bed weight varies between models
  • Adsorbants and secondary amine exchange materials differ between models
Tubes contain ultrapure adsorbants and primary and secondary amine exchange material (PSA). Fluoropolymer resin frits minimize background.
Order Now
SPECIFICATIONS
Environmentally Preferable