Cyclophosphamide monohydrate, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-(Bis(2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide hydrateCyclophosphamide hydrate2-[Bis(2-chloroethyl)amino]tetrahydro-1,3,2-oxazaphosphorin-2-oxide

239785-1GM 239785-1GM
CA80500-080EA 314.76 CAD
80500-080 CA80500-080
Cyclophosphamide monohydrate, Sigma-Aldrich®
Cyclophosphamide monohydrate

A widely used antineoplastic agent that also displays immunosuppressant activity


Induces apoptotic cell death in rat thymus through the Fas/Fas ligand pathway.

Formula: C₇H₁₇Cl₂N₂O₃P
MW: 279.1 g/mol
MDL Number: MFCD00149395
CAS Number: 6055-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR