Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
236108 237105 237107 236105
CA43300-438CS 2137.09 CAD
CA43300-438 CA43300-448 CA43300-450 CA43300-434
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
Black and white 96-well plates are specifically designed for fluorescence and luminescence assays
Nunclon™Δ treated surface ensures optimal cell adhesion and growth. Non-treated surface is ideal for use in liquid phase immunoassays, serological tests and other homogeneous assays. MaxiSorp™ surface has a high affinity for proteins, especially antibodies. PolySorp™ surface is less polar than MaxiSorp and has an affinity towards molecules with mainly hydrophobic domains. MaxiSorp™ and PolySorp™ surfaces are used in many solid phase immunoassays.
Made of polystyrene, they feature a flat-More Product Information