Nunclon™ Delta 96-Well MicroWell™ Plates, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
156545 161093 163320 143761 168055 167008 136101 137101 137103 136102 168136 168055
CA21993-952CS 1890.64 CAD
CA21993-952 CA21993-954 CA25382-329 CA25382-340 CA25382-341 CA25382-342 CA43300-430 CA43300-444 CA43300-446 CA46000-328 CA73521-214 25382-341
Nunclon™ Delta 96-Well MicroWell™ Plates, Sterile, Thermo Scientific
Microplates Cell Culture Plates
These 96-well, polystyrene plates are ideal for cell culture, cloning, viral titration, and cell fusion
The Nunclon™Δ treated surface ensures optimal cell growth and attachment. Plates fit standard automated handling equipment. Plate edges and lids are designed to minimize evaporation. Lids are shaped to facilitate stacking. Raised well rims reduce cross-contamination. White plates provide maximum reflection, minimum autofluorescence and autoluminescence. Black plates provide minimum background and back light scatter in fluorescence assays. Plates also feature alphanumeric well identification and excellent optical quality. Radiation sterilized.
Ordering information: For lids and sealing tapes, see the Nunc® Microplate Lids, Thermo Scientific series.