Synthware Solvent Addition Funnel for Flash Chromatography, Kemtech America
Supplier: Kemtech America
F291275 F291210 F291975 F291910 F292810 F292812
60003-418EA 118.44 CAD
60003-418 60003-420 60003-422 60003-424 60003-426 60003-428
Synthware Solvent Addition Funnel for Flash Chromatography, Kemtech America
Funnels Dropping Funnels
Funnel design permits addition of solvents to flash chromatography columns without disturbing the packing layer.
  • Heavy wall borosilicate construction
  • Graduated stem
The lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Small joint size funnels fit columns with a [ST]24/40 or [ST]35/20 joint. Large joint size funnels fit columns with a large joint such as [ST]45/50 or 50/30 spherical joint. Top funnel diameter: 90mm.