Uvex Ignite™ Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S4401 S4401X S4411X S4413 S4413
CA89041-708EA 15.66 CAD
CA89041-708 CA89041-710 CA89041-712 CA89041-714 CA89041-720 CA89041-722 CA89041-726 CA89041-730 89041-730
Uvex Ignite™ Safety Eyewear, Honeywell Safety
Safety Glasses
Uvex Ignite™ eyewear combines sleek sport-styling and a wraparound design for superior face coverage and protection. Lightweight, frameless design permits 180° of unobstructed and distortion-free vision.
  • Hardcoat and Uvextra® AF antifog coatings provide scratch and chemical resistance
  • Clear lenses are appropriate for most indoor application
  • Silver mirror lenses deflect outdoor sunlight and glare
A comfortable soft saddle pad nosepiece and flexible temple tip pads comfortably secure the eyewear in place and help reduce slip, making these glasses ideal for extended wear. Coatings are permanently bonded to the lenses and will retain their anti-scratch, antistatic, and UV protection properties even after repeated cleanings. Gray lenses minimize outdoor sunlight and glare. SCT-Reflect 50 lenses have a light mirror coating for applications involving varying light conditions.
Certifications: Meets ANSI Z87+ and CSA Z94.3 standards, as well as military Vo ballistic test for impact.