Quick Workup Cartridges for Flash Chromatography, Agela Technologies
Supplier: Phenomenex
QWC003 QWC006 QWH006 QW9001 QW9003 QWS001 QWS003 QWS006 QWA003 QWP003
97009-758PK 1101.48 CAD
97009-758 97009-760 97009-766 97009-768 97009-770 97009-772 97009-774 97009-776 97009-778 97009-782 97009-788
Quick Workup Cartridges for Flash Chromatography, Agela Technologies
Chromatography Columns
These cartridges provide quick work-up of organic synthesis mixture and are ideal for sample cleanup.
Inject reaction mixtures into the cartridge and elute the compounds with appropriate organic solvents.