KONTES® ULTRA-WARE® Safety System 3, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
953930-0000 953930-0001
89005-052EA 644.43 CAD
89005-052 89082-062
KONTES® ULTRA-WARE® Safety System 3, Kimble Chase
Closures Safety Caps and Safety Waste Caps
This cap system eliminates the safety hazards associated with aluminum foil-wrapped solvent containers
The system includes a three-hole PTFE cap, PEEK™ combination inlet filter and sparger, ETFE flangeless nuts and ferrules, ETFE vent port plug, and FEP fluoropolymer resin tubing. Systems are available with or without a solvent bottle adapter that converts GL-45 mobile phase caps to standard 4L bottles for direct connection to an HPLC pump.