(±)-S-Nitroso-N-acetylpenicillamine (SNAP) ≥95% (by HPLC), Calbiochem®

Supplier: MilliporeSigma
Warning

487910-20MG 487910-1SET
CA80053-430EA 346.35 CAD
CA80053-430 CA80501-170
(±)-S-Nitroso-N-acetylpenicillamine (SNAP) ≥95% (by HPLC), Calbiochem®
(±)-S-Nitroso-N-acetylpenicillamine (SNAP)

Nitric oxide donor that begins to evolve nitric oxide immediately upon solubilization in aqueous buffers (t¹/₂ = 10 h). Mimics the actions of nitric oxide, including the relaxation of isolated bovine coronary artery rings (EC50 = 130 nM).


Markedly activates soluble guanylate cyclase. Also reported to cause the reversible inactivation of protein kinase C activity. Induces apoptosis in mouse thymocytes.


Note: 1 set = 4×5 mg.

Formula: C₇H₁₂N₂O₄S
Storage Temperature: Freezer
CAS Number: 79032-48-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR