S-Nitrosoglutathione (GSNO), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione

487920-10MG 487920-50MG 487920-1SET
CA80053-436EA 276.69 CAD
CA80053-436 CA80053-438 CA80503-342
S-Nitrosoglutathione (GSNO), Sigma-Aldrich®
S-Nitrosoglutathione (GSNO)

A utility carrier of nitric oxide. Breaks down to produce nitric oxide and glutathione radical at pH 7.4. Causes smooth muscle relaxation and inhibits platelet aggregation. Induces apoptosis in mouse thymocytes.

Formula: C₁₀H₁₆N₄O₇S
MW: 336.319 g/mol
MDL Number: MFCD00214341
CAS Number: 57564-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR