Pierce™ Sulfo-SBED Biotin Label Transfer Kit and Reagents - Western Blot Application, Thermo Scientific
Supplier: Thermo Scientific
33033 33073
CAPI33033EA 949.98 CAD
CAPI33033 CAPI33073
Pierce™ Sulfo-SBED Biotin Label Transfer Kit and Reagents - Western Blot Application, Thermo Scientific
Protein Linking and Labeling Systems
Sulfo-SBED is a trifunctional crosslinking reagent containing a biotin, a sulfonated N-hydroxysuccinimide (Sulfo-NHS) active ester and a photoactivatable aryl azide
  • Eliminates the need to radiolabel
  • Segregated biotin nucleus
  • Ideal for protein:protein interaction studies using the label transfer method
This allows for easy purification and detection of interacting proteins. This ready-to-use kit combines the most commonly used components in label transfer applications to save preparation time. Components are supplied in single-use packaging to eliminate waste and provide a longer shelf life. In addition, extensive guidelines are provided with the kit to aid in critical aspects of reagent use. For additional purification, add the protein:protein interaction reagent to the protocol. It features a photoreactive aryl nitrene group, a sulfo-NHS-ester group, and a biotin group for increased flexibility in any protocol. The cleavable disulfide bond allows molecules to be easily detached from each other after conjugates are recovered.
Sulfo-SBED Biotin Label Transfer Reagent is a multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.
Delivery information: Label Transfer kit includes Sulfo-SBED 8 x 1 mg, PBS pack, Label Transfer Buffer (20X) 200 mL, Streptavidin-HRP Conjugate 0.1 mg, Dithiothreitol (DTT) 8 x 7.7 mg, Slide-A-Lyzer MINI Dialysis Devices Plus Float 10K MWCO 10-device kit.