Crews® Tacoma® Protective Eyewear, MCR Safety
Supplier: MCR Safety
TC110 TC120 TC110XL
66025-340EA 14.8 CAD
66025-340 66025-344 66025-356
Crews® Tacoma® Protective Eyewear, MCR Safety
Safety Glasses
These durable and economical safety glasses feature a single wraparound lens with sideshields and a browguard.
  • Clear lenses allow 85% light transmission
  • Gray lenses allow 12% light transmission
Glasses have polycarbonate lenses and wire-reinforced plastic temples. The Tacoma™ XL glasses are large glasses designed to fit over prescription lenses.
Certifications: Meet or exceed ANSI Z87.1+, CSA Z94.3, EN166/F, and CEEN standards.